Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trichlorododecylsilane

Catalog No. 44230100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-230-100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 44-230-100ML Supplier Sigma Aldrich Supplier No. 44230100ML
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Trichlorododecylsilane

Specifications

CAS 103128-76-3
Density 0.994 g/mL at 25°C
Boiling Point 72°C (0.4 mmHg, lit.)
Molecular Formula C11H21ClO
Refractive Index n20/D 1.454; n20/D 1.454
Linear Formula C11H21ClO
MDL Number MFCD09039273
Quantity 100 mL
Synonym Dodecyltrichlorosilane
Molecular Weight (g/mol) 204.74 g/mol
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.