Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Chloro-4,4,5,5-Tetramethyl-1,3,2-Dioxaphospholane

Catalog No. 4475361G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-753-61G 1 g
44-753-65G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 44-753-61G Supplier Sigma Aldrich Supplier No. 4475361G
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Chloro-4,4,5,5-tetramethyl-1,3,2-dioxaphospholane (TMDP) can be used:As a reagent for the phosphitylation of alcohols and heteroatomic nucleophiles, resulting in the formation of useful glycosyl donors and ligands.As a phosphitylation reagent to derivatize lignin samples for 31P NMR analysis.

Specifications

CAS 14812-59-0
Density 1.149 g/mL at 25°C
Boiling Point 81.5°C to 82°C (13 mmHg, lit.)
Molecular Formula C6H12ClO2P
Linear Formula C6H12ClO2P
Refractive Index n20/D 1.471
MDL Number MFCD00274239
Quantity 1 g
Molecular Weight (g/mol) 182.59 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.