Learn More
Sigma Aldrich Fine Chemicals Biosciences 45 Methylenedoxy 12 pheny100MG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences D4784
Encompass
4 5-Methylenedioxy-1 2-phenylenediamine dihydrochloride is a fluorescence tag for keto acids. It is also required for the derivatization of sialic acids in presence of dilute sulfuric acid. The method allows determination of sialic acids.
Catalog No. 50-290-6918
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.