Learn More
Mitegen Clear Acrylic 45-Well Microtube Holder, For 1.5mL Tubes, 190mm Dia., Fits Foam Dewars, Enhances Sample Organization
SDP
Supplier:  Mitegen MLCEDTH01
Encompass_Preferred
This Clear Acrylic 45-Well Microtube Holder is designed to optimize laboratory workflow and enhance sample organization, especially within foam dewars. Constructed from premium clear acrylic, it provides unobstructed visibility of your samples, facilitating easy identification and monitoring throughout your experiments. Its 45 precisely-sized 11mm wells securely hold standard 1.5ml microtubes, maximizing storage capacity and ensuring efficient sample handling. With a 190mm diameter, this holder is universally compatible, designed to seamlessly fit on top of most standard foam dewars, providing a snug and stable platform. Durable and easy to clean, it streamlines sample storage and various laboratory workflows involving 1.5ml microtubes. This holder is generally resistant to alkalis, detergents, dilute inorganic acids, and aliphatic hydrocarbons, but should be avoided with esters, ketones, aromatic, or chlorinated hydrocarbons.
Catalog No. NC3627991