Learn More
ABclonal Technology 450 anti-Human CD152/CTLA-4
SDP
Supplier:  ABclonal Technology A26550200T
Encompass_Preferred
This gene is a member of the immunoglobulin superfamily and encodes a protein which transmits an inhibitory signal to T cells The protein contains a V domain a transmembrane domain and a cytoplasmic tail Alternate transcriptional splice variants encoding different isoforms have been characterized The membrane-bound isoform functions as a homodimer interconnected by a disulfide bond while the soluble isoform functions as a monomer Mutations in this gene have been associated with insulin-dependent diabetes mellitus Graves disease Hashimoto thyroiditis celiac disease systemic lupus erythematosus thyroid-associated orbitopathy and other autoimmune diseases
Catalog No. 50-000-11498
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.