Learn More
Abcam 450 nm Stop Solution for TMB Substrate, 100 mL
Description
- Species Reactivity: Species independent
Specifications
Specifications
| Product Type | ELISA Stop Solution |
| Quantity | 100 mL |
| For Use With (Application) | ELISA |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.