Learn More
SHOWA™ 451 Series Natural Rubber Palm Coated Thermal-Insulated Gloves
Catalog No. 19019244 Shop All Showa Best Glove Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-019-244 Medium
19-019-245 Large
19-019-246 X-Large
19-019-243 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-019-244 Supplier SHOWA™ Supplier No. 451M08
Add to Cart
Add to Cart
This item is not returnable. View return policy
Created with warmth in mind, the 451 thermal-insulated gloves are designed to be worn in cold weather.
Benifits
  • Thermal Insulation
  • Flexible
  • Breathable
  • Extra Grip
  • Seamless Knit
  • Skin-friendly
  • Abrasion-resistant
Features
  • Rough Grip
  • Ergonomic
  • Elasticated Wrist Cuff
  • Natural Rubber Palm Coating
Trades & Applications
  • Signalers
  • Brick Laying
  • Carpentry
  • Driving Machines
  • Masonry
  • Demolition
  • Shipping/Receiving
Specifications
Certifications/Compliance CE Cat II, EN 388:2016, EN 511:2006
Coated Area Palm
Construction Seamless Knit
Coating Material Natural Rubber
Color Gray
Material Cotton, Polyester
Size Medium
Liner Material Acrylic
Resistant To Abrasion Resistant
Series 451
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.