Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Polyglycolide

Catalog No. 45762010G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-762-010G 10 g
45-762-01G 1 g
45-762-05G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 45-762-010G Supplier Sigma Aldrich Supplier No. 45762010G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Biodegradable and biocompatible PGA could be potentially useful for bone regeneration. High molecular weight polyglycolide (PGA) can be prepared by ring opening polymerization of diglycolide in the presence of diphenyl bismuth bromide.Polyglycolide (PGA) along with polylactide may be used to prepare a biodegradable copolymer to be used as implantations. Fabrication of biopolymer/carbon nanotube composite has been reported using PGA fiber as one of the constituents of the composite. Modified porous PGA scaffolds has been fabricated.

Specifications

CAS 26124-68-5
Density 1.53 g/mL at 25°C
Linear Formula (C2H4O3)n
MDL Number MFCD00148011
Quantity 10 g
Synonym PGA, Poly(glycolic acid)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.