Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Potassium stannate trihydrate

Catalog No. 462799250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
46-279-9250G 250 g
46-279-91KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 46-279-9250G Supplier Sigma Aldrich Supplier No. 462799250G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Potassium stannate trihydrate

Specifications

Melting Point 140°C
CAS 12125-03-0
Quantity 250 g
Molecular Formula K2O3Sn -+ 3H2O
Linear Formula K2SnO3-+3H2O
MDL Number MFCD00150391
Synonym Potassium tin(IV) oxide
Molecular Weight (g/mol) 298.95
Percent Purity 99.9% trace metals basis
Density 3.197 g/mL (at 25°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.