Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Dimethylaminoethanol

Catalog No. 4714532L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 L
18 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-145-32L 2 L
47-145-3100ML 100 mL
47-145-318L 18 L
47-145-3500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 47-145-32L Supplier Sigma Aldrich Supplier No. 4714532L
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Dimethylaminoethanol

Specifications

CAS 108-01-0
Melting Point -70°C (lit.)
Density 0.886 g/mL (at 20°C (literature))
Boiling Point 134°C to 136°C (lit.)
Molecular Formula C4H11NO
Refractive Index n20/D 1.4294 (literature)
Linear Formula (CH3)2 NCH2CH2OH
MDL Number MFCD00002846
Quantity 2 L
Synonym N,N-Dimethyl-2-hydroxyethylamine; N,N-Dimethylethanolamine; DMEA; Jeffcat DMEA
Molecular Weight (g/mol) 89.14
Percent Purity ≥99.5%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.