Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Bisphenol A propoxylate diglycidyl ether

Catalog No. 475750100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-575-0100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 47-575-0100ML Supplier Sigma Aldrich Supplier No. 475750100ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Bisphenol A propoxylate diglycidyl ether (BPDG) is an epoxy based resin that can be cured by ring opening mechanism of the oxirane groups in the monomer. It can be used to form structurally consistent and thermally stable composites for a variety of applications.BPDG can be used as a reinforcing material for silica aerogel for enhancing the thermo-mechanical properties of the composite. It can also be used to form hydrophilic epoxy networks which can potentially be used for surface coating and structural adhesives.

Specifications

CAS 106100-55-4
MDL Number MFCD01321618
Quantity 100 mL
Synonym Bisphenol A bis(propylene glycol glycidyl ether)ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.