Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly(dimethylsiloxane), bis(3-aminopropyl) terminated

Catalog No. 48168810ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-168-810ML 10 mL
48-168-850ML 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 48-168-810ML Supplier Sigma Aldrich Supplier No. 48168810ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tandem mass spectrometry (MS/MS) reveals that bis(3-aminopropyl)-PDMS mainly dissociates via intramolecular nucleophilic substitutions at the end groups.bis(3-aminopropyl) terminated poly(dimethylsiloxane) may be used in the synthesis of dipicolylamidopropylpoly(dimethylsiloxane) (DPPDMS).

Specifications

CAS 106214-84-0
Density 0.98 g/mL (at 25°C)
Boiling Point >200°C
Linear Formula H2N(CH2)3 Si(CH3)2 O[Si(CH3)2 O]nSi(CH3)2(CH2)3 NH2
Refractive Index n20/D 1.410
MDL Number MFCD00241456
Quantity 10 mL
Synonym PDMS
Molecular Weight (g/mol) 2500
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.