Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Pharmingen™ 488 EdU Click Proliferation Kit, 50 Tests

Catalog No. BDB565455 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB565455 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB565455 Supplier BD Pharmingen™ Supplier No. 565455
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For research use only

The BD Pharmingen™ 488 EdU Click Proliferation Kit provides a tool for analyzing DNA synthesis in cycling cells. In this method, the thymidine analog EdU (5-ethynyl-2'-deoxyuridine) is incorporated into newly synthesized DNA by cells progressing through the S (DNA synthesis) phase of the cell cycle. EdU can then be detected in fixed and permeabilized cells with a fluorophore-labeled azide. The reaction of the fluorophore-labeled azide and EdU occurs via a copper-catalyzed click reaction between the azide moiety of the fluorophore and the alkyne moiety of EdU. When co-stained with a DNA dye such as 7-AAD, PI, or DAPI, cell populations may be segmented by flow cytometry into the G0/G1-phases (2N DNA content, EdU-negative), S-phase (2N-4N DNA content, EdU-positive), or G2/M-phases (4N DNA content, EdU-negative).

The BD Pharmingen™ 488 EdU Click Proliferation Kit contains 6-FAM Azide, which is excited by the blue laser and has an excitation maximum of 496 nm and an emission maximum of 516 nm.

Please note that this kit is provided as Part 1 of 2 (Components A, B, and C) to be stored dry and protected from light at -20°C, and Part 2 of 2 (Components D, E, F, and G) to be stored dry at 2 - 8°C or room temperature.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 50 Tests
For Use With (Equipment) Flow Cytometer
Conjugate Unconjugated
For Use With (Application) Research Use Only
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.