Learn More
Abcam 490/515 Nm Extracellular Vesicle Membrane Staining Kit, 500 Tests
Description
- Assay type: Cell-based
- Detection method: Flow cytometry-fluorescent
- Applications: Flow Cyt
- Sample types: Suspension cells, Adherent cells
- Reactive species: Mammals
- Results type: Quantitative
- Key features: Labeling in < 4 hrs with 30 secs hands-on time; 100% antibody recovery
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Shipping Condition | Ambient |
| Target Molecule | Extracellular Vesicles |
| Reactivity | Mammals |
| Content And Storage | Store at -20°C. |
| For Use With (Equipment) | Flow Cytometer |
| Format | Kit |
| Detection Method | Fluorescence |
| No. of Reactions | 500 Tests |
| Cross Reactivity | Mammals |
| Sample Type | Suspension cells, Adherent cells |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.