Learn More
Description
- Made of B-946 pressure sensitive vinyl withstands elements indoors or out
- Supplied in liner-mounted rolled form
Specifications
Specifications
| Adhesive Type | Acrylic |
| Color | Blue, White |
| Finish | Gloss |
| Material | Vinyl |
| Display Text | 270 Arrows |
| For Use With (Application) | Pipe and Valve Marking |
| Quantity | 1 Ea. |
| Dimensions (L x W) | 30 yd. x 4 in. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.