Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Glow-In-The-Dark Electrical Hazard Signs: DANGER - HIGH VOLTAGE

Catalog No. 19000432 Shop All Brady Corporation Products
Change view
Click to view available options
Dimensions (L x W):
25.4 x 17.7 cm
35.5 x 25.4cm
Material:
Plastic
Polyester
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Material
19-000-432 35.5 x 25.4cm Polyester
19-000-436 25.4 x 17.7 cm Plastic
19-000-437 35.5 x 25.4cm Plastic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-000-432 Supplier Brady™ Supplier No. 80204
Add to Cart
Add to Cart

Photoluminescent sign to identify electrical hazard and high voltage areas

  • Available in Self-Stick Polyester and Plastic materials
  • Legend: HIGH VOLTAGE

Specifications

Material Polyester
Quantity 1 Ea.
Description Glow-In-The-Dark Self-Stick Polyester
Height (English) 10 in.
Height (Metric) 25.4 cm
Width (English) 14 in.
Width (Metric) 35.5 cm
Languages English
Dimensions (L x W) 35.5 x 25.4cm
Displayed Text Header/Legend: DANGER/HIGH VOLTAGE
Product Type Electrical Hazard Sign
Color Red on Glow
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.