Learn More
Brady™ Socks
Catalog No. 19000174 Shop All Brady Corporation Products
Change view
Click to view available options
Packaging Quantity:
12/Cs.
30/Cs.
6/Cs.
Length (English):
4 ft.
8 ft.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity Length (English)
19-000-174 12/Cs. 4 ft.
19-000-176 6/Cs. 8 ft.
19-000-175 30/Cs. 4 ft.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-000-174 Supplier Brady™ Supplier No. AW412
Add to Cart
Add to Cart
Dependable, flexible tubes that contain and absorb liquids
  • Made of all-purpose polypropylene SOC
  • Molds around corners and conforms to uneven surfaces
  • Wringable and can be incinerated
  • Used as a working tool for all major industries
  • Available in variety of types and sizes
Specifications
Material Polypropylene
Type Allwik Socks
For Use With (Application) Industrial applications
Packaging Quantity 12/Cs.
Length (English) 4 ft.
Diameter (English) 3 in.