Learn More
Brady™ Oil Only Socks
Catalog No. 19000183 Shop All Brady Corporation Products
Change view
Click to view available options
Packaging Quantity:
12/Cs.
30/Cs.
4/Cs.
6/Cs.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
19-000-183 12/Cs.
19-000-182 4/Cs.
19-000-185 6/Cs.
19-000-184 30/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-000-183 Supplier Brady™ Supplier No. OIL412
Add to Cart
Add to Cart
Dependable, flexible tubes that contain and absorb liquids
  • Made of oil-only polypropylene SOC
  • Molds around corners and conforms to uneven surfaces
  • Does not absorb water or water-based solutions
  • Wringable and can be incinerated
  • Used as a working tool for all major industries
  • Available in variety of types and sizes
Specifications
Material Polypropylene
Type Oil Only Socks
For Use With (Application) For use against oil and petroleum based spills
Packaging Quantity 12/Cs.