Learn More
Brady™ Re-Form™ Multiwik™ Socks
Catalog No. 19000180 Shop All Brady Corporation Products
Change view
Click to view available options
Packaging Quantity:
4/Cs.
40/Cs.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
19-000-180 4/Cs.
19-000-181 40/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-000-180 Supplier Brady™ Supplier No. MS124
Add to Cart
Add to Cart
Dependable, flexible tubes that contain and absorb liquids
  • Molds around corners, and conform to uneven surface
  • Wringable and incinerable
  • Not for use with aggressive acids and caustics
  • Specifications
    Material Recycled Newsprint, Raw Cellulose
    Type Re-Form Multiwik Socks
    Packaging Quantity 4/Cs.