Learn More
3M™ Scott Fire & Safety™ Industrial SCBA - ISCBA
Catalog No. 19122021
Change view
Click to view available options
Facepiece Type:
AV-2000 (CS) Poly
AV-3000 SureSeal, Dupont™ Kevlar™ aramid fibers, medium
None
Pressure:
2216 psi
4500 psi
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Facepiece Type Pressure
19-122-021 AV-2000 (CS) Poly 2216 psi
19-122-020 None 2216 psi
19-122-023 AV-2000 (CS) Poly 4500 psi
19-122-022 AV-3000 SureSeal, Dupont™ Kevlar™ aramid fibers, medium 2216 psi
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-122-021 Supplier 3M™ Scott Fire & Safety™ Supplier No. IND112013104101
Self-contained breathing apparatus for industrial applications.
  • E-Z Flo mask-mounted pressure demand regulator
  • Features Top Down Convertibility concept, which allows increase in versatility and compatibility with Scott product line
  • Sturdy, zinc-plated carbon steel wire-formed back frame
  • Durable nylon harness system
  • Two-piece polymer waist fastener
  • Alligator-style shoulder clips
  • Audible bell alarm is mounted on first-stage pressure reducer
  • Over-the shoulder luminescent pressure gauge
  • Double-locking latch and durable fabric band allows quick and easy cylinder change
Warranty and Services
Five years
TRUSTED_SUSTAINABILITY
Specifications
Product Type Industrial SCBA
Pressure 2216 psi
Facepiece Type AV-2000 (CS) Poly
Harness Style Nylon polymer buckle
Model IND112013104101
Regulator Type EZ-Flo Standard
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.