Learn More
3M™ Scott Fire & Safety™ Industrial SCBA - ISCBA
Description
- E-Z Flo mask-mounted pressure demand regulator
- Features Top Down Convertibility concept, which allows increase in versatility and compatibility with Scott product line
- Sturdy, zinc-plated carbon steel wire-formed back frame
- Durable nylon harness system
- Two-piece polymer waist fastener
- Alligator-style shoulder clips
- Audible bell alarm is mounted on first-stage pressure reducer
- Over-the shoulder luminescent pressure gauge
- Double-locking latch and durable fabric band allows quick and easy cylinder change
Warranty and Services
Five years
Specifications
Specifications
| Product Type | Industrial SCBA |
| Pressure | 2216 psi |
| Facepiece Type | AV-3000 SureSeal, Dupont™ Kevlar™ aramid fibers, medium |
| Harness Style | Nylon polymer buckle |
| Model | IND112013103101 |
| Regulator Type | EZ-Flo Standard |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.