Learn More
Scott Safety™ ACSi SCBA
Description
- Lightweight, flexible Thermoflex backplate for greater comfort and flexibility
- EZ-Flo regulator reduces inhalation and exhalation resistance for reduced user fatigue
- Pneumatics are easily removed with a Scott “quarter-turn”—no tools required—for ease of maintenance and turn-around
- Adjustable, padded shoulder straps and waistband with quick-release buckle for comfort and ease of donning and doffing
- Shoulder mounted photoluminescent gauge with whistle end-of-service alarm for ease of use
- Cylinder: 2216 psi Aluminum, 30 min.
Specifications
Specifications
| Product Type | Industrial SCBA |
| Pressure | 2216 psi |
| Facepiece Type | AV-2000 (CS) Poly |
| Harness Style | ACSI Standard |
| Hose Type | None |
| Includes | Cylinder: 2216 psi Aluminum, 30 min.; Manifold: Standard |
| Model | ACS3A2013004101 |
| Regulator Type | EZ-Flo Standard |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.