Learn More
Scott Safety™ ACSi SCBA
Catalog No. 19122027
Change view
Click to view available options
Facepiece Type:
AV-2000 (CS) Poly
AV-2000, Dupont™ Kevlar™ aramid fibers
AV-3000 SureSeal (M) Poly
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Facepiece Type
19-122-027 AV-2000, Dupont™ Kevlar™ aramid fibers
19-122-029 AV-3000 SureSeal (M) Poly
19-122-026 AV-2000 (CS) Poly
19-122-025 AV-2000 (CS) Poly
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-122-027 Supplier Scott Safety™ Supplier No. ACS3A2013001101
Offers the Advanced Carrying System (ACS), which features a padded Thermoflex backplate molded to fit the spine and back muscles for increased comfort and flexibility.
  • Lightweight, flexible Thermoflex backplate for greater comfort and flexibility
  • EZ-Flo regulator reduces inhalation and exhalation resistance for reduced user fatigue
  • Pneumatics are easily removed with a Scott “quarter-turn”—no tools required—for ease of maintenance and turn-around
  • Adjustable, padded shoulder straps and waistband with quick-release buckle for comfort and ease of donning and doffing
  • Shoulder mounted photoluminescent gauge with whistle end-of-service alarm for ease of use
  • Cylinder: 2216 psi Aluminum, 30 min.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Industrial SCBA
Pressure 2216 psi
Facepiece Type AV-2000, Dupont™ Kevlar™ aramid fibers
Harness Style ACSI Standard
Hose Type None
Includes Cylinder: 2216 psi Aluminum, 30 min.; Manifold: Standard
Model ACS3A2013001101
Regulator Type EZ-Flo Standard
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.