Learn More
Scott Safety™ AIR-PAK 75i SCBAs
Description
- Streamlined backframe provides comfortable fit with lower profile
- Featues one-piece backframe constructed of lightweight, time-tested, durable aluminum alloy
- Parachute-Style, oversized harness fasteners designed with gloved hands
- Enhanced latching system with durable para-aramid fabric band
- Includes efficient breathing circuit
- Features Top Down Convertibility concept for c increase in versatility, compatibility with Scott product line
- E-Z Flo regulator with Vibralert end-of-service indicator
- Has two dual-redundant pressure reducer interconnected through automatic transfer valve
- Quick-don mounting system in E-Z Flo regulator
- Air-saver switch angled for easy operation with gloved hand
- Fully padded shoulder straps and waist belt
- Integrated donning/carrying handles
- Dual mechanical speaking diaphragms
- Remote pressure gauge
- Cam-Over center cylinder adjustment band
- Drag rescue loop
Specifications
Specifications
| Product Type | SCBA |
| Pressure | 4500 psi |
| Facepiece Type | AV-3000 Sureseal Dupont™ Kevlar™ aramid fibers (M) |
| For Use With (Application) | Industrial applications |
| Includes | Airline option, Hansen HK socket |
| Model | API454002053111 |
| Regulator Type | E-Flo regulator w/Vibralert Quick Connect |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.