Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,4-Butanediol

Catalog No. 49373218L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 L
3 L
18 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
49-373-218L 18 L
49-373-21L 1 L
49-373-23L 3 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 49-373-218L Supplier Sigma Aldrich Supplier No. 49373218L
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1,4-Butanediol

Specifications

CAS 110-63-4
Melting Point 16°C
Density 1.017 g/mL at 25°C
Boiling Point 230°C
Molecular Formula C4H10O2
Refractive Index n20/D 1.445
Linear Formula HO(CH2)4OH
MDL Number MFCD00002968
Quantity 18 L
Synonym 1,4-Butylene glycol; Tetramethylene glycol
Molecular Weight (g/mol) 90.12 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.