Learn More
MilliporeSigma™ Novagen™ HT96™ NovaBlue Competent Cells
Catalog No. 710114MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Plate
4 Plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-011-4MI 4 Plates
71-011-3MI 1 Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 71-011-4MI Supplier MilliporeSigma™ Supplier No. 710114
Add to Cart
Add to Cart
Includes
HT96 NovaBlue Competent Cells (1 or 4 plates), SOC Medium (1 x 14mL or 4 x 14mL), Test Plasmid (1 x 10μL or 2 x 10 μL), 8-Cap Strips (1 or 4), Reagent Reservoirs (1 or 4), Plate Lids (1 or 4)
High-efficiency competent cells predispensed in 96-well plate for high-throughput applications
NovaBlue is a K-12 strain ideally suited as an initial cloning host because it has high transformation efficiency, blue/white screening capability (with appropriate plasmids) and mutations in recA and endA which result in high yields of high-quality plasmid DNA.
  • Cells dispensed in 20μL volumes in 96 well polypropylene plate compatible with thermal cyclers and water baths
  • Wells are individually sealed and have raised rims to prevent cross-contamination
  • Seals can be easily pierced with standard pipet tips or removed for access
  • Strips of caps are also provided for reliable sealing during manipulation and storage
  • Groups of 24 wells can be easily split from the plate for processing smaller numbers of samples
  • Efficiency: >1 × 108CFU/μg
TRUSTED_SUSTAINABILITY
Specifications
Efficiency > 1 x 108CFU/μg
Content And Storage Store HT96 Competent Cells, SOC Medium and Test Plasmid at -70°C or below. Store Cap Strips, Plate Lids and Reagent Reservoirs at room temperature
For Use With (Application) For high-throughput transformation
Quantity 4 Plates