Description
- Equipped with inert and non-corrosive plunger
- Fitted with resistant ultra-high molecular weight poly-ethylene (UHMWPE) plunger tip
Applications:
For life science applications with salty buffers, mining applications with acids or trace metal analysis
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Product Type | Gastight Saltline Syringe |
| Volume (Metric) | 1 mL |
| Connection Type | Bubble Free Prime (BFP) |