Learn More
Cellpath FiloSlide Slide Storage Packs
SDP
Catalog No. 22222054
Change view
Click to view available options
Type:
FiloSlide 4 Pk. w/100 Wallets
FiloSlide 5 Pk. w/100 Wallets
FiloSlide 6 Pk. w/150 Wallets
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
22-222-054 FiloSlide 5 Pk. w/100 Wallets
22-222-055 FiloSlide 6 Pk. w/150 Wallets
22-222-051 FiloSlide 4 Pk. w/100 Wallets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-222-054 Supplier Cellpath Supplier No. WED990000A
Item Discontinued This product is either new or inactive, and cannot be ordered online.
FiloSlide cabinet with plastic slide wallets
  • Superior slide protection
  • Document wallet on reverse to store paperwork
  • FiloSlide 4, 5, 6 Wallets made from clear flexible plastic
  • Ideal for building reference/teaching sets of slides
  • Up to 150 FiloSlide 6 can be stored in FiloSlide cabinet
FiloSlide 4 slide wallet:
  • Four microscope slide pockets, one document pocket measuring 103 x 153mm (4 x 6 in.)
  • Combination pack of 100 wallets and one FiloSlide cabinet also available
FiloSlide 5 slide wallet:
  • Five microscope slide pockets, one document pocket
  • Combination pack of 100 wallets and one FiloSlide cabinet is also available
FiloSlide 6 Slide wallet:
  • Six microscope slide pockets, one document pocket measuring 220 x 150mm (8.6 x 5.9 in.) and one index pocket
  • Each pack of 10 FiloSlide 6 wallets is supplied with 50 color coded index cards (10 each of red, blue, green, white, yellow)
  • Combination pack of 150 wallets and one FiloSlide cabinet is available
Specifications
Type FiloSlide 5 Pk. w/100 Wallets
For Use With (Application) Building reference/teaching sets of slides
Includes 100 plastic slide wallets; 5 microscope slides pockets; 1 document pocket (103 x 153m