Learn More
Brady™ SXT Oil Only Heavy Rolls
Catalog No. 17111181 Shop All Brady Corporation Products
Change view
Click to view available options
Capacity (English):
18 gal./Bale
37 gal.
Dimensions (L x W):
150 ft. x 15 in.
150 ft. x 30 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Dimensions (L x W)
17-111-181 18 gal./Bale 150 ft. x 15 in.
17-111-188 37 gal. 150 ft. x 30 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-111-181 Supplier Brady™ Supplier No. SXT15P
Add to Cart
Add to Cart
Oil only sorbent made to handle petroleum-based spills
  • Remains intact even when saturated
  • Spunbond coverstock on both sides
  • Ideal for workstations involving the wipe down of tools or component parts
  • Perforated
  • Specifications
    Capacity (English) 18 gal./Bale
    Material Polypropylene
    Type Oil Only Heavy Roll
    For Use With (Application) Food Processing, High Tech Industries, Machining of small parts, Paper Industry, Pharmaceutical
    Packaging Quantity 1 Ea.
    Weight Capacity Heavyweight
    Dimensions (L x W) 150 ft. x 15 in.