Learn More
Brady™ Oil Only Drag Nets
Catalog No. 17111065 Shop All Brady Corporation Products
Change view
Click to view available options
Includes:
30 nets on 50 ft. rope
30 nets, no rope
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
17-111-065 30 nets on 50 ft. rope
17-111-118 30 nets, no rope
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-111-065 Supplier Brady™ Supplier No. ODN08
Floats in water and entraps heavy oil
  • Made up of flat, chemical-resistant filaments in half-pound units
  • Can be re-used after draining or dousing
  • Extracted oil can be recycled or downgraded as fuel oil
  • Used as sweep between two boats or deployed along shoreline
  • Available in two sizes: 30 nets on 50 ft. rope and 30 nets (no rope)
Specifications
Type Oil/Drag Net
Includes 30 nets on 50 ft. rope