Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Spill Pallet Drums

Catalog No. 17111282 Shop All Brady Corporation Products
Change view
Click to view available options
Dimensions (L x W x H):
52 x 26 x 17 in.
52 x 52 x 17 in.
Holds:
2 Drums
4 Drums
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H) Holds
17-111-282 52 x 26 x 17 in. 2 Drums
17-111-283 52 x 52 x 17 in. 4 Drums
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-111-282 Supplier Brady™ Supplier No. SCDP2
Add to Cart
Add to Cart

For containing potentially hazardous leaks

  • Injection molding construction
  • Includes grate pins to prevent the walls from bulging
  • Interchangeable grades with large openings for easy leak detection and liquid removal
  • Nestable and forklift friendly
  • Available in 4-drum and 2-drum configurations
  • Specifications

    Capacity (English) 68.7 gal.
    Capacity (Metric) 260.6 L
    Material Polyethylene
    Type 2-drum Spill Pallet
    Holds 2 Drums
    Dimensions (L x W x H) 52 x 26 x 17 in.
    Weight (English) 46.48 lb.
    Weight (Metric) 102.49 kg
    WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.