Learn More
Brady™ Cansorb™ Oil Only Granular Sorbents
Catalog No. 17111078 Shop All Brady Corporation Products
Change view
Click to view available options
Capacity (English):
10 gal.
20 gal.
Type:
Compressed Granular Sorbent
Loose Granular Sorbent
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Type
17-111-078 20 gal. Compressed Granular Sorbent
17-111-077 10 gal. Loose Granular Sorbent
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-111-078 Supplier Brady™ Supplier No. CSCB18
Ideal for oil-only clean up on open water and land applications
  • Made from fiber sphagnum peat moss
  • Class: Oil only
  • Absorbs 8 times its weight
  • Available in compressed and loose granular format
  • Specifications
    Capacity (English) 20 gal.
    Material Fiber Sphagnum Peat Moss
    Type Compressed Granular Sorbent
    Size 18