Learn More
DuPont™ Tychem™ 6000 Hooded Coverall
Catalog No. 19162807 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Medium
3X-Large
4X-Large
5X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-162-807 3X-Large
19-162-810 5X-Large
19-162-808 4X-Large
19-162-813 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-162-807 Supplier DuPont™ Supplier No. TF145TGY3X0006TV
Add to Cart
Add to Cart
This item is not returnable. View return policy
Widely used by military personnel and first responders for chemical warfare situations
  • Zipper features a semi-automatic locking slider allowing the puller to stay down and in a locked position
  • Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing.
  • Three-piece hood design with longer zipper that extends to chin for complete coverage of neck area and good fit around a respirator
  • Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
  • Elastic opening for tighter fit at wrist
  • Elastic opening for tighter fit at ankle
  • Manufactured under specifications that do not contain natural rubber latex
  • Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks.
Compliance
TAA compliant
TRUSTED_SUSTAINABILITY
Specifications
Size 3X-Large
Gender Unisex
Material Tychem
Color Gray
Ankle Style Elastic
Closure Type Zipper
Seam Type Taped