Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Biohazard Identification Signs: Biohazard Infectious Agent Entry Requirements Contact Phone

Catalog No. 19140983 Shop All Brady Corporation Products
Change view
Click to view available options
Material:
Aluminum
Plastic
Polyester
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-140-983 Polyester
19-140-987 Plastic
19-140-993 Aluminum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-140-983 Supplier Brady™ Supplier No. 126647

For identification of equipment, rooms and materials that contain, or are contaminated with, hazardous agents

  • Made from aluminum, plastic or self-sticking polyester
  • Size: 14 in. H x 10 in. W
  • Legend: BIOHAZARD INFECTIOUS AGENT___ ENTRY REQUIREMENTS ___ CONTACT ___ PHONE ___

Specifications

Material Polyester
Description Self-sticking Polyester
Height (English) 14 in.
Height (Metric) 4.2 m
Width (English) 10 in.
Width (Metric) 3 m
Displayed Text BIOHAZARD INFECTIOUS AGENT___ ENTRY REQUIREMENTS ___ CONTACT ___ PHONE ___
Product Type Biohazard Identification Sign

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.