Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Miller™ BackBiter Tie-Back Lanyards

Catalog No. 17990510 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Leg Configuration:
Double-Leg
Single-leg
Includes:
1 locking snap hook, 1 5K snap hooks
1 locking snap hook, 2 5K snap hooks
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Leg Configuration Includes
17-990-510 Single-leg 1 locking snap hook, 1 5K snap hooks
17-990-509 Double-Leg 1 locking snap hook, 2 5K snap hooks
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-990-510 Supplier Honeywell™ Supplier No. 940B/6FTGN
  • $948.00
    $871.65 / Each of 1

    Save $76.35

    8% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Features all-in-one lanyard with SofStop Shock Absorber and cross-arm anchorage connector

  • Innovative BackBiter tie-back lanyard with a 5K Snap Hook
  • Designed from top to bottom for tie-back use
  • Has locking snap and 5K snap hook
  • Contains Double SofStop pack for 3.7m (12 ft.) free fall/duty-rated 400 lb. capacity for up to 1.8m (6 ft.) free fall
  • Available in 6 ft. single and double leg
  • Recommended industries/use: Construction, manufacturing, steel and metals, transportation
  • Hazards: Confined space, fall from height

Certifications

OSHA - 1910.66, 1926.104, 1926.502, ANSI - A10.32, Z359.13, CSA - Z259.11 Class E4, Z259.11 Class E6

Specifications

Leg Configuration Single-leg
Load Bearing Capacity (Metric) 181.4 kg
Load Bearing Capacity (English) 400 lb.
Certifications/Compliance OSHA - 1910.66, 1926.104, 1926.502, ANSI - A10.32, Z359.13, CSA - Z259.11 Class E4, Z259.11 Class E6
Includes 1 locking snap hook, 1 5K snap hooks
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.