Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Excelta™ Medical Tubing Flush Cutter

Catalog No. 18100980 Shop All Excelta Corporation Products
Change view
Click to view available options
Capacity (English):
0.13 in.
0.25 in.
0.38 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
18-100-980 0.38 in.
18-100-979 0.13 in.
18-100-977 0.25 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-100-980 Supplier Excelta™ Supplier No. 292238
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Allows clean cut to variety of soft tubing materials

Allows clean cut to variety of soft tubing materials

  • ESD safe, ergonomic, soft cushioned grips
  • Replaceable blades
  • Made of stainless steel
  • Has dual leaf springs that provide smooth action
  • Available in different cutting capacity: 6.35mm (0.25 in.), 3.3mm (0.13 in.), 9.6mm (0.38 in.) tubing

Specifications

Capacity (English) 0.38 in.
Cut Type Flush
Grip Type Soft-cushioned
Includes w/Replacement blade
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.