Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Excelta™ Medium Stainless Steel Pliers

Catalog No. 18100948 Shop All Excelta Corporation Products
Change view
Click to view available options
Product Type:
Chain-Nose Pliers
Needle nose plier
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
18-100-948 Chain-Nose Pliers
18-100-949 Needle nose plier
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-100-948 Supplier Excelta™ Supplier No. 2844LD
Add to Cart
Add to Cart

Has dual leaf springs for smooth action

  • Available in Chain/Needle nose shape
  • ESD safe, soft-cushioned foam grip
  • Has serrated jaws, long handles
  • Dimension: L x Wx H: 150 x 10.63 x 6.5mm (6 x 0.425 x 0.26 in.)
  • Diffrent Tip Size: 0.88 x 1.38mm (0.035 x 0.055 in.), 0.63 x 1.13mm (0.025 x .045 in.)

Specifications

Product Type Chain-Nose Pliers
Length (English) 6 in.
Length (Metric) 150 mm
Material Stainless Steel
Grip Type Soft-cushioned foam
Jaw Type Serrated
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.