Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Excelta™ Serrated Locking Hemostat

Catalog No. 18100967 Shop All Excelta Corporation Products
Change view
Click to view available options
Tip Style:
Curved
Straight
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Tip Style
18-100-967 Straight
18-100-969 Curved
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-100-967 Supplier Excelta™ Supplier No. 39SE
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Available in Straight/Curved shape

  • Clean room safe
  • Made of stainless steel
  • Overall length: 200mm (8 in.)

Specifications

Product Type Locking hemostat
Length (English) 8 in.
Length (Metric) 200 mm
Material Stainless Steel
Tip Style Straight
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.