Learn More
Justrite™ Floor Grating w/Sump Liner
Description
- For use with 2-,4-,6-,9-,12-,16-drum lockers
- Made of High density polyethylene (HDPE) sump liner
- Fiberglass grating
Applications:
For acid and corrosive chemical spill resistance applications
Specifications
Specifications
| Product Type | Floor Grating |
| Material | HDPE |
| Description | HDPE sump liner |
| For Use With (Equipment) | 9-drum/ (296 cu. ft.) lockers |
| Includes | Sump liner |