Description
- For use with 6-,9-,12-,16-drum fire-rated Agri-Turf™ lockers only
- Available in 4.4kWh (15000 BTU) and 5.9kWh (20000 BTU) energy capacity
Applications:
Adds air conditioning where comfort, chemical stability or freeze protection is important
†For optimum temperature control, appropriate size R-11 insulation package must be ordered when unit is purchased in conjunction with fire-rated models. This accessory must be purchased in conjunction with locker to ensure proper installation and to retain FM-approval.
Specifications
Specifications
| Product Type | Explosion-proof Air conditioner |
| For Use With (Equipment) | 6-,9-drum fire rated Agri-Turf lockers |