Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KleenGuard™ G35 Seamless Knit Nylon Inspection Gloves

Catalog No. 19167726 Shop All Ansell Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-167-726 X-Small
19-167-727 Small
19-167-728 Medium
19-167-729 Large
19-167-730 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-167-726 Supplier KleenGuard™ Supplier No. 38716
Add to Cart
Add to Cart
This item is not returnable. View return policy

Low-lint gloves are ideal for detecting imperfections and defects when conducting critical quality control inspections. Ambidextrous KleenGuard™ G35 Seamless Knit Nylon Inspection Gloves are silicone-free for use in a wide variety of applications including film handling, aerospace, and automotive assembly.

  • Ambidextrous design allows gloves to be replaced one at a time, rather than in pairs
  • High degree of sensitivity and dexterity for inspecting for defects and imperfections
  • Designed for critical quality control inspections
  • Seamless knit nylon shell reduces skin irritation
  • Low lint for cleanroom applications
  • Silicone-free
TRUSTED_SUSTAINABILITY

Specifications

Construction Seamless Knit
Coating Material Uncoated
Color White
Disposable Single-use
Hand Ambidextrous
Latex Free Yes
Material Nylon
Product Type Inspection Gloves
Size X-Small
Sterility Non-sterile
For Use With (Application) Automotive Assembly, Aerospace Engineering, and Quality Control Inspections
Glove Cuff Style Knit
Series KleenGuard™ G35
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.