Learn More
Spectrum™ Labs Spectra/Por™ 7 1000 D MWCO Standard RC Pre-Treated Dialysis Kits
Shop All Spectrum Medical Industries Products

Description
Includes
- 1meter of pre-treated tubing, a pair of closures and 5 opening picks
- Hydrophilic, symmetric porositiy
- Equilibrated in water
- Packaged wet in preservative solution of 0.05% sodium azide
- For small experiments or qualifying a membrane for a particular application
Specifications
Specifications
| MWCO | 1 kDa |
| Diameter (Metric) | 11.5 mm |
| Length (Metric) | 1.1 mL/cm vol./length |
| Quantity | 1 m Tubing/Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.