Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tricyclo[5.2.1.02,6]decanedimethanol diacrylate

Catalog No. 496669100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
49-666-9100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 49-666-9100ML Supplier Sigma Aldrich Supplier No. 496669100ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tricyclo[5.2.1.02,6]decanedimethanol diacrylate is an acrylate monomer that has low shrinkage and high refractive index. It can be used in the synthesis of shape memory polymers for the fabrication of optoelectronic and stretchable electronic devices. It can also be used in the formation of holographic polymer dispersed liquid crystals (HPDLCs).

Specifications

CAS 42594-17-2
Density 1.1 g/mL (at 25°C)
Molecular Formula C18H24O4
Refractive Index n20/D 1.506 (literature)
Linear Formula C18H24O4
Quantity 100 mL
Synonym Bis(acryloyloxymethyl)tricyclo[5.2.1.02,6 ]decane; Dicyclopentyldimethylene diacrylate; Dimethyloldicyclopentane diacrylate; Dimethyloltricyclodecane diacrylate; Tricyclo[5.2.1.02,6 ]decanedimethanol diacrylate; Tricyclodecanedimethanol diacrylate
Molecular Weight (g/mol) 304.38
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.