Learn More
Sigma Organic Chemistry Lithium | 100G | 7439-93-2 | MFCD00134051 | 99% trace metals basis
Supplier: Sigma Organic Chemistry 499811100G
Lithium, 100G
About This Item:
Storage: room temp
EINECS Number: 231-102-5
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.