Learn More
US Plastic Corporation 4 oz. Wide Mouth French Square Glass Bottles with 33/400 P Caps, Case of 24
SDP
Supplier:  US Plastic Corporation 72484
Encompass
This clear space saver is appropriate for powders and liquids. Choose glass bottles if you need zero permeation containers exposed to temperature extremes, you need rigidity, you value product integrity, you want to eliminate waste, or you are concerned about the environment.P = black phenolic polycone lined capSquare shape provides for easy pouring & fillingDimensions: 2.073" Dia. x 4.390" Hgt.Sold in full cases only
Catalog No. NC3454920