Learn More
Biocare Medical, LLC 4PLUS AP 1000 UNIVERSAL 100ML
SDP
Supplier:  Biocare Medical, LLC AP506US
Encompass_Preferred
4plus AP Universal Detection is a two-step detection designed for sensitivity and reliability. This detects either mouse or rabbit primary antibodies with a biotinylated secondary antibody followed by an Alkaline Phosphatase (AP) label.
Catalog No. 50-823-64
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.