Learn More
Biocare Medical, LLC 4plus Streptavidin HRP Label, 25ml
SDP
Supplier:  Biocare Medical, LLC HP604H
Encompass_Preferred
4plus Streptavidin HRP Label is the second step in a two-step detection designed for sensitivity and reliability. This detects a biotinylated secondary antibody with an Streptavidin-Horseradish Peroxidase (HRP) conjugate.
Catalog No. 50-828-80
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.