Learn More
REPLIGEN CORPORATION Repligen ACBT-500-S1S, 500 mL Max Volume, Irradiated Conical Reservoir
SDP
Supplier:  REPLIGEN CORPORATION ACBT500S1S
Encompass
The Repligen ACBT-500-S1S is a 500 mL irradiated conical reservoir designed for efficiency in critical fluid management. It offers a maximum working volume of 500 mL, and a low hold-up volume of less than 10 mL, ensuring optimal fluid recovery. With materials engineered for compatibility and sterility, it supports various laboratory applications and bioprocessing workflows, making it a reliable choice for researchers.
Catalog No. NC1990770
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.