Learn More
Electron Microscopy Sciences Electropolished Stainless Steel Freezer Rack 4x4, Holds 16 3" boxes
Supplier: Electron Microscopy Sciences 6161525
High quality, ultra-durable freezer racks for long term sample storage. All edges have been ground down to remove the sharp surfaces that plague standard freezer racks. Electropolishing cleans up all weld points that, over time, may be the starting points for corrosion and leaves racks ultra-shiny and free from any surface contaminants. Superior construction and design have created a more sturdy rack than what is currently available on the market. Racks are designed and finished to last longer in the demanding freezer environment where samples are housed.
Capacity: Holds 16 3" boxes
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.