Learn More
USA Scientific Inc 5.0 mL TipOne Repeat Dispens
Supplier:  USA Scientific Inc 47610500
Encompass
5.0 mL TipOne positive displacement tip for repeating pipettes, str, individually wrapped, 100 tips
Catalog No. 50-274-5294
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.